Learn More
Bochem™ Stainless Steel Wire Basket, Square
Description
- Stainless steel
- Square
- Flat bottom for stability
Specifications
Specifications
| Dimensions (L x W) | 250 L x 150 Wmm |
| Dimensions (L x W x H) | 250 x 150 x 150 mm |
| Weight (Metric) | 410 g |
| Product Type | Wire Basket |
| Array | 250 mm |