Learn More
Bochem™ Stainless Steel Wire Basket, Square, 2 Cross-ties
Product Code. 10234271
Change view
Click to view available options
Dimensions (L x W x H):
400x300x200
500 x 300 x 250 mm
600 x 400 x 300 mm
Unit Size:
Each
Product Code. Dimensions (L x W x H) unitSize
10234271 600 x 400 x 300 mm Each
10576371 400x300x200 Each
10692112 500 x 300 x 250 mm Each
3 options
Product Code. 10234271
Brand: Bochem™ 10076
Only null left
Add to basket
Add to basket
Place laboratory items in the Bochem™ Stainless Steel Wire Basket, Square, 2 Cross-ties. .
  • Mesh width 8x8x1mm,
  • Square
  • Stainless Steel
  • 2 cross-ties
Specifications
Dimensions (L x W) 600 L x 400 Wmm
Dimensions (L x W x H) 600 x 400 x 300 mm
Product Type Wire Basket
Array 600 mm