Learn More
Ohaus™ Stainless Steel Weight, OIML F1, OH
Product Code. 15683384 Shop All Ohaus Products
Change view
Click to view available options
Weight (Metric):
100 g
200 g
1 kg
Unit Size:
Each
Product Code. Weight (Metric) unitSize
15683384 200 g Each
15673384 100 g Each
15742854 1 kg Each
3 options
Product Code. 15683384
Brand: Ohaus™ 11117042
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Single body design weight adjusted to meet or exceed 0IML F1 tolerances
  • Stainless steel construction
  • Include a weight case
  • Single body design without adjusting cavity
TRUSTED_SUSTAINABILITY
Specifications
Description Weight, 200g, OIML F1, OH
Classification OIML F1 Weights
Material Stainless Steel
Weight (Metric) 200 g
Tolerance Meets or exceeds OIML F1