Learn More
Testo™ Stainless Steel Waterproof Food Probe
Click to view available options
Response Time:
7 sec.
Unit Size:
Each
Specifications
Specifications
| Includes | Food Probe, 4 dia. x 125mm L Probe Shaft, 3.2 dia. x 50mm L Probe Shaft Tip, Stainless Steel, –60 to 400°C Measuring Range, 7 sec. Response Time, Waterproof |
| Certifications/Compliance | EN 60584-2 |
| Material | Stainless Steel |
| Response Time | 7 sec. |