Learn More
S Murray and Co™ Stainless Steel Watchmaker Forceps
Product Code. 12740926
Change view
Click to view available options
Size:
No. 4
No. 7
Unit Size:
Each
Product Code. Size unitSize
12740926 No. 7 Each
12311379 No. 4 Each
2 options
Product Code. 12740926
Brand: S Murray and Co™ E010/07
Only null left
Add to basket
Add to basket
Use these forceps for dissection and manipulating slides and cover glasses. S Murray™ Stainless Steel Watchmaker Forceps are made from high grade stainless steel in accordance to the specifications of ISO 7153-1.
  • Stainless Steel forceps
  • For dissection
Specifications
Product Type Forceps, Watchmaker
Material Stainless Steel
For Use With (Application) For dissection
Quantity Each
Size No. 7
Length (Metric) 110 mm