Learn More
S Murray and Co™ Stainless Steel Watchmaker Forceps
Product Code. 12311379
Change view
Click to view available options
Size:
No. 4
No. 7
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12311379 No. 4 Each
12740926 No. 7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12311379 Supplier S Murray and Co™ Supplier No. E010/04
Add to basket
Add to basket
Use these forceps for dissection and manipulating slides and cover glasses. S Murray™ Stainless Steel Watchmaker Forceps are made from high grade stainless steel in accordance to the specifications of ISO 7153-1.
  • Stainless Steel forceps
  • For dissection
Specifications
Product Type Forceps, Watchmaker
Material Stainless Steel
For Use With (Application) For dissection
Quantity Each
Size No. 4
Length (Metric) 110 mm