Learn More
Buerkle™ Stainless Steel Valve Lance Samplers
Product Code. p-8011254 Shop All Bürkle Products
Click to view available options
Length (Metric):
550 mm
800 mm
Unit Size:
Each
Product Code. 11951571
Brand: Buerkle™ 53591600
Only null left
Add to basket
Add to basket
Sampler for all-layer samples of viscous, paste-like, semi-liquid substances, according to DIN 51750 pt. 3 and DIN EN ISO 15528. Different layers remain intact particularly with paste-like substances.
  • Stainless steel AISI 316L (1.4404)
  • 32 mm Ø
  • Optimised opening mechanism (can be completely disassembled) for easy cleaning∣
TRUSTED_SUSTAINABILITY
Specifications
Product Type Valve Lance Sampler
Material Stainless Steel
For Use With (Application) All-Layer Samples of Viscous, Paste-Like, Semi-Liquid Substances, According to DIN 51750 pt. 3 and DIN EN ISO 15528.
Diameter (Metric) 32 mm
Length (Metric) 550 mm