Learn More
Thermo Scientific™ Stainless Steel Unions, Tees and Crosses
Product Code. 10715146 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cross
Tee
Union
Description:
0.010 in. through hole
0.020 in. through hole
0.050 in. through hole
0.062 in. through hole
Swept Volume:
∼0.0 μL
0.025 μL
0.103 μL
0.129 μL
0.134 μL
0.57 μL
0.72 μL
0.836 μL
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Description Swept Volume Unit Size
10715146 Tee 0.020 in. through hole 0.57 μL Each
10107384 Union 0.020 in. through hole 0.103 μL Each
10669365 Union 0.020 in. through hole 0.134 μL Each
10046364 Union 0.020 in. through hole 0.129 μL Each
10302985 Cross 0.020 in. through hole 0.72 μL Each
10097214 Union 0.010 in. through hole 0.025 μL Each
10007224 Union 0.050 in. through hole 0.836 μL Each
10189004 Union 0.062 in. through hole ∼0.0 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10715146 Supplier Thermo Scientific™ Supplier No. U428
Estimated Shipment: 23-11-2026
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
  • Absolute zero or low dead volume formats
  • Includes two stainless steel nuts and ferrules
TRUSTED_SUSTAINABILITY
Specifications
Type Tee
Swept Volume 0.57 μL
Includes Tee
Description 0.020 in. through hole
Material Stainless Steel
For Use With (Equipment) 1/16 in. OD Tubing
Length (Metric) 22.61 mm