Description
- Made from 304 stainless steel
- Available in a range of different lengths
- Robust design
- Useful for handling intricate items such as tissues
Specifications
Specifications
| Tip Style | Serrated |
| Length (English) | 15.7 in. |
| Material | Stainless Steel (304) |
| Length (Metric) | 400 mm |
| Product Type | Tweezers |
| Quantity (No. per Case) | 1 Tweezer/Cs. |
| Sterility | Non-sterile |
| Style | Standard |