Learn More
Cole-Parmer™ Stainless Steel Tweezers
Product Code. 11725449 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
11.43 cm
Unit Size:
Each
Product Code. Length (Metric) unitSize
11725449 11.43 cm Each
1 options
Product Code. 11725449
Brand: Cole-Parmer™ 0728702
Only null left
Add to basket
Add to basket
Recommended for micro electronics and precision assembly
  • Finely crafted stainless steel tweezers
  • Feature hand finished points for accuracy and excellent tip hardness
  • Anti-glare finish reduces eye strain
  • Anti-magnetic, anti-acid and acid resistant
Specifications
Tip Style Spate Tip
Length (English) 4.75 in.
Material Stainless Steel
Length (Metric) 11.43 cm
Product Type Tweezers
Type Straight Tweezers