Learn More
Bochem™ Stainless Steel Tubing Clips
Product Code. 10010751
Change view
Click to view available options
Diameter (Metric):
10 mm
12 mm
14 mm
25 mm
29 mm
33 mm
Unit Size:
Each
Product Code. Diameter (Metric) unitSize
10010751 12 mm Each
10798111 10 mm Each
10306101 25 mm Each
10664032 33 mm Each
10316101 29 mm Each
10020751 14 mm Each
6 options
Product Code. 10010751
Brand: Bochem™ 12131
Only null left
Add to basket
Add to basket
Used to control the flow of liquids. Bochem™ Stainless Steel Tubing Clips are manufactured from stainless steel.
  • Manufactured from stainless steel
  • Used with tubing
Specifications
Diameter (Metric) 12 mm
Material Stainless Steel
Product Type Tubing Clip