Learn More
Ohaus™ Stainless Steel Tube Holder
Shop All Ohaus Products
Click to view available options
Holds:
1.5 to 2 mL Microtubes
Unit Size:
Each
Description
- OHAUS offers a large selection of accessories that can be used on all OHAUS vortex mixers
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Specifications
| Holds | 1.5 to 2 mL Microtubes |
| Product Type | Tube Holder |
| Material | Stainless Steel |
| Description | 1.5-2.0 mL Stainless Steel Tube Holder |