Learn More
Agencinox™ Stainless Steel Trolleys
Product Code. 11789503
Change view
Click to view available options
Dimensions (L x W):
600 x 400 mm
600 x 430 mm
750 x 500 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11789503 750 x 500 mm Each
11789493 600 x 400 mm Each
11769503 600 x 430 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11789503 Supplier Agencinox™ Supplier No. 745
  • ONLINE EXCLUSIVE PRICE
    6675.95 NOK
    3182.74 NOK / Pack of 1
    Save 3493.21 NOK
    52% Off
View all promotions
Estimated Shipment: 23-12-2026
Use these trolleys for a variety of applications. Agencinox™ Stainless Steel Trolleys are made of 18/10 stainless steel with bright polish finishing.
  • Heavy duty structure
  • Easy cleaning
  • With 2 handles, shaped trays
  • Frame: 18/10 stainless steel tube diam. 25mm
  • Castors diam. 75mm
Specifications
Dimensions (L x W) 750 x 500 mm
Product Type Trolley
Includes Shelves
Load Bearing Capacity (Metric) 100 kg
Material 18/10 Stainless Steel
No. of Shelves 2