Learn More
S Murray and Co™ Stainless Steel Treve's Rat Tooth Forceps
Product Code. 12710926
Change view
Click to view available options
Length (Metric):
125 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12710926 125 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12710926 Supplier S Murray and Co™ Supplier No. E062/01
  • ONLINE EXCLUSIVE PRICE
    36.65 NOK
    30.64 NOK / Pack of 1
    Save 6.01 NOK
    16% Off
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Treve's Rat Tooth Forceps are ideal for major and minor surgery sets.
  • Stainless Steel
  • Length: 125mm
Specifications
Product Type Treve's Rat Tooth Forceps
Material Stainless Steel
For Use With (Application) For dissection
Quantity Each
Length (Metric) 125 mm