Learn More
Amsta Rostfritt Stainless Steel Transport Flask - Acid Proof Churn with Lid
Product Code. p-200113433
Click to view available options
Capacity (Metric):
8 L
10 L
Unit Size:
Pack of 10
Product Code. 30285000
Brand: Amsta Rostfritt 51632101
Highest quality stainless steel transport flask - acid proof. Made of 1 mm stainless steel (EN1.4404). Intended for antibiotics, vaccines, chemicals and liquids. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
Specifications
Product Type Pail
Capacity (Metric) 10 L
Color Silver
Material Stainless Steel (316L)