Learn More
Bochem™ Stainless Steel Transport Container with Lid and Handle
Brand: Bochem™ 8304
Additional Details : Weight : 0.10000kg
Specifications
Transport Container | |
Stainless Steel (18/10) | |
240 mm |
5 L | |
180 mm | |
Lid and Handle |