Learn More
Amsta Rostfritt Stainless Steel Thermo Canteen Insulated With Lid
Product Code. p-200113438
Change view
Click to view available options
Capacity (Metric):
10 L
15 L
20 L
25 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
30284865 10 L Each
30285022 15 L Each
30284894 20 L Each
30284925 25 L Each
4 options
Product Code. 30284865
Brand: Amsta Rostfritt 50675101
Highest quality stainless steel thermo canteen. Manufactured in Sweden.
  • Stainless steel 304 /1.4301
Specifications
Product Type Pail
Capacity (Metric) 10 L
Color Silver
Material Stainless Steel