Learn More
RETSCH Stainless Steel Test Sieves Intermediate Rings
Product Code. p-8004997 Shop All Retsch Products
Change view
Click to view available options
Size:
100 mm dia. x 40 mm H
200 mm dia. x 25 mm H
200 mm dia. x 50 mm H
203 mm dia. x 25 mm H
203 mm dia. x 50 mm H
305 mm dia. x 40 mm H
400 mm dia. x 65 mm H
450 mm dia. x 100 mm H
Unit Size:
Each
Product Code. Size unitSize
10156981 100 mm dia. x 40 mm H Each
10496431 200 mm dia. x 50 mm H Each
11944422 203 mm dia. x 50 mm H Each
10564112 200 mm dia. x 25 mm H Each
11984422 203 mm dia. x 25 mm H Each
10790473 305 mm dia. x 40 mm H Each
11964432 450 mm dia. x 100 mm H Each
10563922 400 mm dia. x 65 mm H Each
8 options
Product Code. 10156981
Brand: RETSCH 60.935.000100
Only null left
Specifications
Product Type Test Sieve Intermediate Ring
Material Stainless Steel
Diameter (Metric) 100 mm
Height (Metric) 40 mm
For Use With (Equipment) Vibratory Sieve Shaker AS 200 basic
Size 100 mm dia. x 40 mm H