Learn More
RETSCH Stainless Steel Test Sieves Intermediate Rings
Shop All Retsch Products
Click to view available options
Size:
100 mm dia. x 40 mm H
200 mm dia. x 25 mm H
200 mm dia. x 50 mm H
203 mm dia. x 25 mm H
203 mm dia. x 50 mm H
305 mm dia. x 40 mm H
400 mm dia. x 65 mm H
450 mm dia. x 100 mm H
Unit Size:
Each
Specifications
Specifications
| Product Type | Test Sieve Intermediate Ring |
| Material | Stainless Steel |
| Diameter (Metric) | 203 mm |
| Height (Metric) | 50 mm |
| For Use With (Equipment) | Vibratory Sieve Shaker AS 200 basic |
| Size | 203 mm dia. x 50 mm H |