Specifications
| Product Type | Test Sieve Intermediate Ring |
| Material | Stainless Steel |
| Diameter (Metric) | 200 mm |
| Height (Metric) | 25 mm |
| For Use With (Equipment) | Vibratory Sieve Shaker AS 200 basic |
| Size | 200 mm dia. x 25 mm H |