Learn More
Fisherbrand™ Stainless Steel Tank, 38L
Product Code. 17245556 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
38 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17245556 38 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17245556
Estimated Shipment: 01-12-2026
For water bath.
For use with Fisherbrand™ Digital Heating Circulator.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 38 L
Dimensions (D x W x H) 410 x 860 x 410 mm