Learn More
VACUUBRAND™ Stainless Steel T-Pieces With Small KF Flanges
Product Code. 10682222
Change view
Click to view available options
Diameter (Metric) Inside:
10/10/10 mm
16/16/16 mm
25/25/25 mm
40/40/40 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Unit Size
10682222 40/40/40 mm Each
10450463 16/16/16 mm Each
10059570 10/10/10 mm Each
10009600 25/25/25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10682222 Supplier VACUUBRAND™ Supplier No. 20673516
Estimated Shipment: 12-01-2027
Specifications
Diameter (Metric) Inside 40/40/40 mm
For Use With Solenoid-operated valves VV and VV C
Material Stainless Steel
Quantity 1 Each
Size 65 x 130 mm
Type T-piece