Learn More
Swann-Morton™ Stainless Steel Surgical Scalpel Handle B3
missing translation for 'orderingAttributeHoverText'
Size:
# 3
# 4
Handle Type:
Barron Handle
Long Handle
missing translation for 'unitSize'
Each
Pack of 10
Description
- Ergonomic design
- Precise, finely crafted
- Suitable for a variety of procedures and blades
Specifications
Specifications
| Blade Material | Stainless Steel |
| Product Type | Scalpel Handle |
| Handle Type | Long Handle |
| Size | # 4 |
| For Use With (Equipment) | Blade # 18, # 19, # 20, # 21, # 22,# 22A, # 23, # 24,# 25, # 25A, # 26, # 27 and # 36 |
| Sterility | Non-sterile |
| Capacity (Metric) | 4 L |