Learn More
Swann-Morton™ Stainless Steel Surgical Scalpel Handle B3
Description
- Ergonomic design
- Precise, finely crafted
- Suitable for a variety of procedures and blades
Specifications
Specifications
| Blade Material | Stainless Steel |
| Product Type | Scalpel Handle |
| Handle Type | Long Handle |
| Size | # 3 |
| For Use With (Equipment) | Blade # 6, # 9, # 10, # 10A, # 11, # 11P, # 11E, # 12, # 12D, # 13, # 14, # 15, # 15A, # 15C, # 15D, # 15T, # 16, # 40 and # SG3. |
| Sterility | Non-sterile |
| Capacity (Metric) | 3 L |