Learn More
Usbeck Laborgeraete™ Stainless Steel Support Rods
Product Code. 10739932
Change view
Click to view available options
Length (Metric):
250 mm
450 mm
1000 mm
Thread Size:
M10
Without Thread
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Thread Size Unit Size
10739932 1000 mm M10 Each
10629522 1000 mm Without Thread Each
10300562 250 mm M10 Each
10239101 450 mm Without Thread Each
10167391 450 mm M10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10739932 Supplier Usbeck Laborgeraete™ Supplier No. 2144
  • ONLINE EXCLUSIVE PRICE
    261.00 NOK
    240.30 NOK / Pack of 1
    Save 20.70 NOK
    7% Off
 Restricted Item
Estimated Shipment: 07-01-2027
Specifications
Material Stainless Steel (18/10)
Length (Metric) 1000 mm
Diameter (Metric) Inner 12 mm
Product Type Support Rod
Thread Size M10
Quantity 1 Ea.