Description
- Constructed of stainless steel
- 450mm in length
Specifications
Specifications
| Diameter (Metric) | 10 mm |
| For Use With (Equipment) | Magnetic Stirrer |
| Includes | M8 Fixing Nut |
| Length (Metric) | 450 mm |
| Type | Rod |