Learn More
Cole-Parmer™ Stainless Steel Straight Union Compression Fitting
Click to view available options
For Use With:
Fits 1/2 in. Tubing O.D.
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel |
| For Use With | Fits 1/2 in. Tubing O.D. |
| Min. Operating Temperature (Metric) | -253°C |
| Packaging Quantity | 1 Pack |
| Max. Operating Temperature (Metric) | 648°C |
| Product Type | Straight Union Compression Fitting |