Learn More
S Murray and Co™ Stainless Steel Storkbill Spring Forceps
Product Code. 12700926
Change view
Click to view available options
Length (Metric):
110 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12700926 110 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12700926 Supplier S Murray and Co™ Supplier No. E017/01
  • ONLINE EXCLUSIVE PRICE
    66.85 NOK
    55.89 NOK / Pack of 1
    Save 10.96 NOK
    16% Off
Estimated Shipment: 18-12-2026
Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Storkbill Spring Forceps are ideal for major and minor surgery sets.
Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Storkbill Spring Forceps are ideal for major and minor surgery sets.
  • Stainless Steel
  • Length: 10mm
Specifications
Product Type Storkbill Spring Forceps
Material Stainless Steel
For Use With (Application) For dissection
Quantity Each
Length (Metric) 110 mm