Learn More
Interscience Laboratories BagMixer™ Paddle Blender Bag Storage
Description
- Stackable.
- Autoclavable.
- All stainless steel.
- BagRack™ allows clean and methodical storage of bags which can be left open to pipet easily or can be closed with a BagClip™.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Holds | Blood Bag Holders (12) |
| Material | Stainless Steel |
| Shape | Rectangular |
| Stackable | Yes |
| For Use With (Application) | For 12 Bags 400 mL |
| Quantity | 1 Ea. |
| Product Type | Storage Rack |