Learn More
Agencinox™ Stainless Steel Sterilization Boxes
Product Code. 11713944
Change view
Click to view available options
Height (Metric):
40 mm
50 mm
60 mm
70 mm
100 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
11713944 100 mm Each
11703924 40 mm Each
11783934 50 mm Each
11793934 60 mm Each
11703944 70 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11713944 Supplier Agencinox™ Supplier No. 5075
Only null left
Add to basket
Add to basket
Use these boxes for a variety of general laboratory applications. Agencinox™ Stainless Steel Sterilization Boxes keep instruments safe and secure during sterilization process.
  • Stainless steel instrument boxes
  • Available in a variety of sizes
Specifications
Material Stainless Steel
Shape Rectangular
Product Type Sterilization Box
Length (Metric) 450 mm
Width (Metric) 200 mm
Height (Metric) 100 mm