Learn More
Bochem™ Stainless Steel Stand Base
Product Code. 10363621
Change view
Click to view available options
Length (Metric):
210 mm
250 mm
300 mm
315 mm
Width (Metric):
130 mm
150 mm
160 mm
200 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
10363621 300 mm 150 mm Each
10281921 210 mm 130 mm Each
10752542 250 mm 160 mm Each
10527561 315 mm 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10363621 Supplier Bochem™ Supplier No. 5002
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 08-12-2026
Support your supplies with the Bochem™ Stainless Steel Stand Base, which has a reinforcing leg for stability.
  • Expanded, flat and heavy feet for stability
  • 1 adjustable foot
  • Anti-slide rubber protection
  • Thread M10
Specifications
Material Stainless Steel (18/10)
Height (Metric) 3 mm
Length (Metric) 300 mm
Width (Metric) 150 mm
Product Type Stand Base
Quantity 1 Pk.