Learn More
Hirschmann™ Stainless Steel Stand
Product Code. 12631845
Change view
Click to view available options
Length (Metric):
500 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12631845 500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12631845 Supplier Hirschmann™ Supplier No. 9317001
  • ONLINE EXCLUSIVE PRICE
    2625.00 NOK
    1661.23 NOK / Pack of 1
    Save 963.77 NOK
    36% Off
Estimated Shipment: 15-12-2026
Provides support in titration applications.
  • Accessory item
  • 500 mm long
  • Manufactured from stainless steel
Specifications
For Use With (Equipment) Titration Apparatus
Type Stand
For Use With (Application) Provides support
Length (Metric) 500 mm