Learn More
S Murray and Co™ Stainless Steel Spring Forceps
Product Code. 12391369
Change view
Click to view available options
Length (Metric):
115 mm
125 mm
150 mm
Tip Style:
Blunt, Narrow Flutes
Sharp, Curved, Narrow Flutes
Sharp, Narrow Flutes
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tip Style Unit Size
12391369 125 mm Blunt, Narrow Flutes Each
12381369 125 mm Sharp, Curved, Narrow Flutes Each
11340705 115 mm Sharp, Narrow Flutes Each
12371369 125 mm Sharp, Narrow Flutes Each
12700046 150 mm Sharp, Narrow Flutes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12391369 Supplier S Murray and Co™ Supplier No. E003/02
Estimated Shipment: 29-12-2026
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
  • Stainless Steel forceps for dissection
Specifications
Product Type Spring Forceps
Material Stainless Steel
For Use With (Application) For dissection
Quantity Each
Tip Style Blunt, Narrow Flutes
Length (Metric) 125 mm