Learn More
Bel-Art™ Stainless Steel Spoons
Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
0.5 mL, 2 mL
0.8 mL, 2.8 mL
1.6 mL, 3 mL
1.8 mL, 5.0 mL
2.0 mL, 8.0 mL
Length (Metric):
15 cm
18 cm
21 cm
25 cm
30 cm
Unit Size:
Pack of 2
Description
Double-ended with two bowl sizes.
- Made from 304 stainless-steel
- Sampling spoons have solid, rounded shaft
- Available in plain polished stainless-steel or coated with Fluo-Kem™ PTFE FEP
Specifications
Specifications
| Autoclavable | Autoclavable |
| Shape | Solid, Rounded Shaft |
| Capacity (Metric) | 1.8 mL, 5.0 mL |
| Product Type | Spoon |
| Finish | Plain-polished 304 grade stainless-steel |
| Material | Stainless Steel |
| Length (Metric) | 25 cm |