Description
Double-ended with two bowl sizes.
- Made from 304 stainless-steel
- Sampling spoons have solid, rounded shaft
- Available in plain polished stainless-steel or coated with Fluo-Kem™ PTFE FEP
Specifications
Specifications
| Autoclavable | Autoclavable |
| Shape | Solid, Rounded Shaft |
| Capacity (Metric) | 0.8 mL, 2.8 mL |
| Product Type | Spoon |
| Finish | Plain-polished 304 grade stainless-steel |
| Material | Stainless Steel |
| Length (Metric) | 18 cm |