Learn More
Bel-Art™ Stainless Steel Spoons
Product Code. 11726545 Shop All SP Scienceware Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL, 2 mL
0.8 mL, 2.8 mL
1.6 mL, 3 mL
1.8 mL, 5.0 mL
2.0 mL, 8.0 mL
Length (Metric):
15 cm
18 cm
21 cm
25 cm
30 cm
Unit Size:
Pack of 2
Product Code. Capacity (Metric) Length (Metric) unitSize
11726545 2.0 mL, 8.0 mL 30 cm Pack of 2
15550122 0.8 mL, 2.8 mL 18 cm Pack of 2
15570122 1.8 mL, 5.0 mL 25 cm Pack of 2
11706545 0.5 mL, 2 mL 15 cm Pack of 2
11716545 1.6 mL, 3 mL 21 cm Pack of 2
5 options
Product Code. 11726545
Brand: Bel-Art™ F367300030
Only null left
Add to basket
Add to basket
Double-ended with two bowl sizes.
  • Made from 304 stainless-steel
  • Sampling spoons have solid, rounded shaft
  • Available in plain polished stainless-steel or coated with Fluo-Kem™ PTFE FEP
Specifications
Autoclavable Autoclavable
Shape Solid, Rounded Shaft
Capacity (Metric) 2.0 mL, 8.0 mL
Product Type Spoon
Finish Fluo-Kem Teflon FEP coated ends
Material Stainless Steel
Length (Metric) 30 cm