Learn More
Buerkle™ Stainless Steel Spatula
Product Code. 11971551 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
180 mm
300 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
11971551 300 mm Each
11878502 180 mm Each
2 options
Product Code. 11971551
Brand: Buerkle™ 53130300
Only null left
Add to basket
Add to basket
Universal instrument for collecting, dividing and mixing pastes, creams etc.
Universal instrument for collecting, dividing and mixing pastes, creams etc.
  • Blade made of stainless steel V2A (1.4301)
  • Wooden handle
TRUSTED_SUSTAINABILITY
Specifications
Product Type Spatula
For Use With (Application) Universal Instrument for Collecting, Dividing and Mixing Pastes, Creams
Blade Material Stainless Steel V2A
Handle Material Wood
Material Stainless Steel Blade, Wood Handle
Length (Metric) 300 mm