Learn More
asecos™ Stainless Steel Small Parts Basket
Artikelnummer. 17846556 Alle ansehen asecos Produkte
Ansicht ändern
Click to view available options
Capacity (English):
58.12 gal.
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Capacity (English) unitSize
17846556 58.12 gal. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17846556 Lieferant asecos™ Lieferanten-Nr. 40450
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
The Stainless Steel Small Parts Basket is useful for small parts cleaner with capacity of 8 litres.
  • Can be hooked onto the upper part of the small parts cleaner to drain off the parts.
  • For small parts cleaner with capacity of 8 litre.s
  • Dia x H (mm): 222
Spezifikation
Product Type Parts Cleaner
Capacity (English) 58.12 gal.
Capacity (Metric) 220 L
Color Blue
For Use With (Application) Safe storage of water pollutans
For Use With (Equipment) 4 x 200 L drum
Material Stainless Steel
Includes PE-grid
Quantity 1
Length (Metric) 1330 mm
Width (English) 52.36 in.
Width (Metric) 1330 mm
Height (English) 12.60 in.
Height (Metric) 320 mm
Weight (English) 121.25 lb.
Weight (Metric) 55 kg
Mehr anzeigen Weniger anzeigen