Learn More
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill
Product Code. 10134133 Shop All Kinematica Products
Click to view available options
Pore Size:
0.2 mm
0.5 mm
0.8 mm
1 mm
1.5 mm
2.0 mm
3 mm
4 mm
5 mm
6 mm
Unit Size:
Each
Product Code. 10134133
Brand: Kinematica™ 35095015
Only null left
Add to basket
Add to basket
Depending on the sample, even finer results can be achieved by grinding with two or more sieves with progressively smaller hole sizes.
Specifications
Product Type Sieve
Material Stainless Steel
Pore Size 1.5 mm