Learn More
Ohaus™ Stainless Steel Shot
Product Code. 15702599 Shop All Ohaus Products
Click to view available options
Type:
Shot
Unit Size:
Each
Product Code. 15702599
Brand: Ohaus™ 30400178
Only null left
Add to basket
Add to basket
OHAUS™ Stainless Steel Shot is suitable for use with all OHAUS™ Dry Block Heaters.
  • For use with modular blocks
  • Constructed of stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Description Stainless Steel Shot, 0.5 kg (1 Lb)
Material Stainless Steel
Type Shot
Weight (English) 1 lb.
Weight (Metric) 0.5 kg