Description
- Stainless steel AISI 316L (1.4404)
- Electro-polished
- Total length 1000 mm
Specifications
Specifications
| Product Type | Scratcher |
| Material | Stainless Steel (V4A) |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 25 mL |
| For Use With (Application) | Designed for Sampling Cohesive, Caked and Not Free-Flowing Substances |
| Length (Metric) | 1000 mm |