Learn More
Amsta Rostfritt Stainless Steel Scoops
Product Code. p-8009090
Change view
Click to view available options
Volume (Metric):
1 L
1.5 L
2 L
Unit Size:
Each
Product Code. Volume (Metric) unitSize
11791826 1 L Each
11701836 1.5 L Each
11731776 2 L Each
3 options
Product Code. 11791826
Brand: Amsta Rostfritt 517551
Only null left
Add to basket
Add to basket
Buckets manufactured to handle chemicals, etc. Made of 0.8 mm acid-resistant stainless steel plate (EN1.4404).
Fully welded with soldered handles without slips for dirt hides.
Specifications
Product Type Scoop
Material Stainless Steel
Volume (Metric) 1 L