Learn More
Rogo Sampaic™ Stainless Steel Scissors
Product Code. 11738531
Change view
Click to view available options
Length (Metric):
140 mm
Tip Style:
Round/Round
For Use With (Application):
Anatomical
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tip Style For Use With (Application) Unit Size
11738531 140 mm Round/Round Anatomical Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11738531 Supplier Rogo Sampaic™ Supplier No. FMN015
  • ONLINE EXCLUSIVE PRICE
    335.00 NOK
    215.36 NOK / Pack of 10
    Save 119.64 NOK
    35% Off
Estimated Shipment: 11-12-2026
Excellent for a variety of cutting applications in the laboratory. Rogo Sampaic™ Stainless Steel Scissors have a beautiful polished matte finish.
  • Surgical grade stainless steel
  • Available in a variety of styles - Metzenbaum, dissecting, iris, anatomical
Specifications
Product Type Stainless Steel Scissors
Length (Metric) 140 mm
Material Stainless Steel
For Use With (Application) Anatomical
Tip Style Round/Round