Learn More
Swann-Morton™ Metal Diecast Handle, Postmortem Scalpel
Product Code. 12922555
Change view
Click to view available options
Blade Material:
Stainless Steel
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Blade Material Unit Size
12922555 Stainless Steel Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12922555 Supplier Swann-Morton™ Supplier No. 2652
  • ONLINE EXCLUSIVE PRICE
    1340.00 NOK
    1211.25 NOK / Pack of 10
    Save 128.75 NOK
    9% Off
View all promotions
Estimated Shipment: 25-01-2027
Use the Swann Morton™ Metal Diecast Handle, Postmortem Scalpel for postmortem surgical techniques.
  • Heavy duty carbon steel blade
  • Metal handle with screw-type fitting
Specifications
Blade Material Stainless Steel
Product Type Handle
For Use With (Equipment) PM40 or PM40 Bull Nose Pathology Blade
Description Epoxy Powder Coated