Learn More
Gestigkeit Harry™ Stainless Steel Sand Bath
Product Code. 10519511
Change view
Click to view available options
Temperature Range:
30°C to 110°C
370°C
50°C to 300°C
Dimensions (D x W x H):
358 x 358 x 230 mm
364 x 514 x 230 mm
Unit Size:
Each
Product Code. Temperature Range Dimensions (D x W x H) unitSize
10519511 50°C to 300°C 358 x 358 x 230 mm Each
10272771 30°C to 110°C 364 x 514 x 230 mm Each
10391272 50°C to 300°C 364 x 514 x 230 mm Each
10614741 370°C 358 x 358 x 230 mm Each
10634741 370°C 364 x 514 x 230 mm Each
5 options
Product Code. 10519511
Brand: Gestigkeit Harry™ ST72