Description
- Handles made of solid stainless steel
- Seals made of PTFE
- High degree of chemical resistance
- Save space
Specifications
Specifications
| Closure Type | Screw cap |
| Capacity (Metric) | 5 L |
| Color | Silver |
| Material | Stainless Steel |
| Includes | Screw Cap, Pressure Relief Valve |
| Thickness (Metric) | 0.8 mm |
| Dimensions | 200 x 130 x 310 mm |
| Length (Metric) | 200 mm |
| Width (Metric) | 130 mm |
| Height (Metric) | 310 mm |
| Show More |