Learn More
Buerkle™ Stainless Steel Safety Jerrycans
Product Code. 11951551 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
Includes:
Fine Dosage Spout, Pressure-relief Valve
Fine Dosage Spout, Pressure-relief Valve, Ventilation
Screw Cap
Screw Cap, Pressure Relief Valve
Unit Size:
Each
Product Code. 11951551
Brand: Buerkle™ 26017010
Only null left
Add to basket
Add to basket
Buerkle™ Stainless Steel Safety Jerrycans offer safety through flame protection and excess pressure valve.
  • Handles made of solid stainless steel
  • Seals made of PTFE
  • High degree of chemical resistance
  • Save space
TRUSTED_SUSTAINABILITY
Specifications
Product Type Safety Jerrican
Capacity (Metric) 10 L
For Use With (Application) Transport
Material Stainless Steel
Includes Screw Cap
Shape Square
Thickness (Metric) 0.6 mm
Dimensions 280 x 130 x 380 mm
Length (Metric) 280 mm
Width (Metric) 130 mm
Height (Metric) 380 mm