Learn More
Buerkle™ Stainless Steel Safety Jerrycans
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
Includes:
Fine Dosage Spout, Pressure-relief Valve
Fine Dosage Spout, Pressure-relief Valve, Ventilation
Screw Cap
Screw Cap, Pressure Relief Valve
Unit Size:
Each
Description
- Handles made of solid stainless steel
- Seals made of PTFE
- High degree of chemical resistance
- Save space
Specifications
Specifications
| Closure Type | Screw cap |
| Capacity (Metric) | 10 L |
| Color | Silver |
| Material | Stainless Steel |
| Includes | Screw Cap, Pressure Relief Valve |
| Thickness (Metric) | 0.8 mm |
| Dimensions | 280 x 130 x 380 mm |
| Length (Metric) | 280 mm |
| Width (Metric) | 130 mm |
| Height (Metric) | 380 mm |
| Show More |