Learn More
Buerkle™ Stainless Steel Safety Container
Product Code. p-7230480 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
10 L
25 L
50 L
Closure Type:
Screw Cap, Pressure-relief Valve
Tab, Pressure-relief Valve
Tap, Pressure Relief Valve
For Use With (Application):
Storage
Storage and Transfer
Unit Size:
Each
Product Code. 15360605
Brand: Buerkle™ 26014010
Only null left
Add to basket
Add to basket
Buerkle™ Stainless Steel Safety Container offer safety through flame protection and excess pressure valve.
  • Safety containers for storage made of high-quality stainless steel AISI 304 (1.4301) / AISI 316 Ti (1.4571) with thickness of sides of 0.8mm
  • Protection from exploding through flame protection and excess pressure valve, seals made of PTFE, the containers have a high degree of chemical resistance
  • Pressure balance takes place from about 0.3 bar
  • Constant ventilation can be set via a plastic lever on the screw cap
TRUSTED_SUSTAINABILITY
Specifications
Product Type Safety Container
Closure Type Tap, Pressure Relief Valve
Capacity (Metric) 10 L
For Use With (Application) Storage and Transfer
Material Stainless Steel
Includes Tab, Pressure-relief Valve, Contents Display
Thickness (Metric) 0.8 mm
Height (Metric) 350 mm
Diameter (Metric) Outer 260 mm