Learn More
asecos™ Stainless Steel Safety Canister
Product Code. 17876536 Shop All asecos Products
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
Height (Metric):
169.00 mm
241.00 mm
271.00 mm
280.00 mm
470.00 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) unitSize
17876536 10 L 280.00 mm Each
17826476 5 L 169.00 mm Each
17806506 10 L 241.00 mm Each
17816406 20 L 271.00 mm Each
17846486 20 L 470.00 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17876536 Supplier asecos™ Supplier No. 36484
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
  • Incl. multilingual safety sticker for content labelling, with filling tap made of stainless steel (1.4301)
  • Incl. ventilation, NBR seals and aluminium screw cap integrated flame arrester and plastic protective ring
Specifications
Product Type Safety Can
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Color Gray
For Use With (Application) safe distribution and dosing of flammable liquids
Material Stainless Steel
Quantity 1
Width (English) 6.69 in.
Width (Metric) 170.00 mm
Height (English) 11.02 in.
Height (Metric) 280.00 mm
Weight (English) 6.61 lb.
Weight (Metric) 3 kg